Alverine citrate salt, powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0215473225 0215473205
ICNA0215473225EA 371 EUR
ICNA0215473225 ICNA0215473205
Alverine citrate salt, powder
Alverine citrate salt

Used for studies of pharmacological activity including: synergism with fluconazole against Saccharomyces cerevisiae and Candida albicans, efficacy of combination with simeticone on abdominal pain associated with irritable bowel syndrome, nutrient-sensitised screening for drugs that shift energy metabolism from mitochondrial respiration to glycolysis,drug effects viewed from a signal transduction network perspective, enhanced cytotoxic effect of proteasome inhibitor MG132, mechanism of action on phasic smooth muscles.

Formule: C₂₆H₃₅NO₇
MDL Nummer: MFCD00035086
CAS nummer: 5560-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR