Mineral Mixture, AIN-76, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0290545502 0290545510
ICNA0290545502EA 93.9 EUR
ICNA0290545502 ICNA0290545510
Mineral Mixture, AIN-76, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
AIN-76 mineral mixtures are components of an animal diet.
  • Mixture is available in powdered form
For complete diet composition, please refer to the product datasheet.