Cleaning solution, concentrate, 7X™, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
097667093 097667094 097667095 097667098 0976670A4
ICNA097667093EA 67.2 EUR
ICNA097667093 ICNA097667094 ICNA097667095 ICNA097667098 ICNA0976670A4
Cleaning solution, concentrate, 7X™, MP Biomedicals
Reinigingsmiddelen Detergenten en oppervlaktereinigers
Safe and effective cleaning solution for glass and plastic labware.
  • Bluish cloudy liquid completely soluble in water at any concentration
  • Drains from glassware in seconds
  • Contains powerful sequestering reagents
  • Ideal for cell cultureware and general labware
The 7X™ laboratory detergent is ideal for thorough cleaning of cultureware and general lab equipment including test tubes, Petri dishes, culture flasks and cylinders, burettes, pipettes, slide covers, and hypodermic syringes. A 5% solution in distilled water has a pH range of 6 to 9. This neutralMore Product Information