Quantus®, Underwear T-Shirts
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
QNTS151-5UWBU0XSIDEA 0 EUR
QNTS151-5UWBU0XSID 113-9182 113-9181 113-9184 113-9183 113-9185
Quantus®, Underwear T-Shirts
Werkkleding
The Quantus® underwear T-shirt is made from a breathable polyester microfibre material with carbon. The fabric was specially developed for use as underwear in combination with a cleanroom overall or coat. It serves as a prefilter to allow open-pore-fabric to be used in a higher cleanroom class.
  • T-shirt (half arm) with round neck and bordered cuffs
  • Extra material flap for bar code inside
  • Soft grip and greater comfort when wearing
  • Anti-static and antimicrobial protection
  • Recommended for wear underneath disposable and reusable clothing
  • RFID chip included
  • Washing instructions: max 60 °C
Weight: 97,5 g/m²
Bestelinformatie: Non standard sizes are available on request. Please contact your local Avantor office for further information.