Cryoboxes, Cardboard
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TE22240 TE22230
479-0697EA 6.13 EUR
479-0697 479-0695
Cryoboxes, Cardboard
Dozen Dozen voor cryobuisjes
Cryoboxes, cardboard, white, for storage of cryogenic samples.
  • Made of water repellant paper
  • Long durability
  • Space saving
  • Easy to assemble
The dividers are 100% free from debris and free from dust.
Leveringsinformatie: Supplied with 9×9 cell dividers.