Cleaning solution for washing machines, liquid concentrate, 7X-O-Matic®, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
097667493 097667494 097667495
ICNA097667493EA 109 EUR
ICNA097667493 ICNA097667494 ICNA097667495
Cleaning solution for washing machines, liquid concentrate, 7X-O-Matic®, MP Biomedicals
Reinigingsmiddelen Detergenten en oppervlaktereinigers
The 7X-O-Matic® solution is a low foaming concentrated surfactant and is particularly suited for use in automatic and mechanical washers.
  • Alkaline, clear, light yellow solution
  • Soluble in water at any concentration
  • Solutions have alkaline salts, such as complex phosphates, added as builders
  • Ideal for cell cultureware and general labware
  • Cleans both glass and plastic labware
Solution contains a powerful wetting agent which will clean microscopic crevices not usually accessible to ordinary detergents. Effective at temperatures ranging from ambient to 85 °C. Solution drains completely from glassware in only a few seconds. A 1% solution in distilled water has a pH range of 9 to 11.