Visuele kleurenkits, chloridetest, MQuant®, Supelco®
Leverancier: MerckTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Quick tests, visual colour comparison.
- Results in approximately 2 minutes
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® titrimetric and colorimetric methods
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
Leveringsinformatie: Elke testkit bevat alle benodigde reagentia, reactiecuvetten, een kleurvergelijkingskaart en uitgebreide gebruiksinstructies.