Transfer buffer, Tris-glycine,10X, AccuGENE™
Leverancier: Lonza
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
733-1629EA 213 EUR
733-1629
Transfer buffer, Tris-glycine,10X, AccuGENE™
Buffers Buffers voor elektroforese Transferbuffers
AccuGENE™ electrophoresis buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Ready to use solutions
  • Reliable manufactured according to strict quality control standards to ensure lot to lot consistency
  • Guaranteed DNase-, RNase- and protease-free
Each reagent is prepared with 18 megOhm water, filtered using a 0,2 micron filter, and filled into sterile bottles.