VWR® Ficoll® 400 Biotechnology Grade
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E965-50GEA 198 EUR
E965-50G E965-100G
VWR® Ficoll® 400 Biotechnology Grade
Media voor celscheiding
Ficoll® 400 is a neutral, hydrophilic polymer of sucrose, which readily dissolves in aqueous solutions.
Ficoll® is a registered trademark of GE Healthcare group of companies.