Ficoll® 400
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
21373.01 21373.02 39763.01 39763.02
SERA21373.01EA 0 EUR
SERA21373.01 SERA21373.02 SERA39763.01 SERA39763.02
Ficoll® 400
Media voor celscheiding
Ficoll® 400 is a neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution. Concentrations up to 50% (w/v), covering a density range up to 1,2 g/ml, can be obtained without exceeding normal osmolarity. Ficoll™ 400 is suitable for the preparation of continuous and discontinuous density gradients, for stabilisation of membrane-bound particles, and for isolation of lymphocytes.
  • Better osmotic properties than sucrose, preserving functional and morphological integrity
  • Does not penetrate biological membranes
  • Readily soluble in aqueous solution
  • Molecular biology grade (free from DNase and RNase) is suitable for use in hybridisation buffers like Denhardt's Solution
Ficoll® is a registered trademark of Pharmacia Biotech AB.