Pierce™ Immunoassay Microplates, 96-Well Polystyrene Plates
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15041 15042
PIER15041EA 534 EUR
PIER15041 PIER15042
Pierce™ Immunoassay Microplates, 96-Well Polystyrene Plates
Microplaten Immunologie-microplaten en -strips
PS, treated to produce high-capacity antibody-binding characteristics for immunoassay applications.
  • Plates are certified for performance, having consistently low coefficients of variation among wells
  • Hydrophobic-ionic binding interaction
  • Corner notch for use in standard plate readers
Clear plates for colorimetric ELISA and other assays. White opaque plates for fluorescent or chemiluminescent assays.
Order Now
Specificaties
Environmentally Preferable