Pierce™ Immunoassay Microplates, Antibody Coated, 96 Well
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15134 15135
PIER15134EA 267 EUR
PIER15134 PIER15135
Pierce™ Immunoassay Microplates, Antibody Coated, 96 Well
Microplaten Immunologie-microplaten en -strips
Pierce™ Antibody coated plates are ideal for binding assays when available antibodies are in low quantities or they denature and become inactive upon direct adsorption to polystyrene plates. Clear plates for colorimetric ELISA and other assays. White plates for chemiluminescent. Black plates for fluorescent assays.
  • Antibody-binding capacity is higher than direct adsorption onto polystyrene
  • Ideal for direct, indirect, competitive or sandwich assays
  • Activation level: 100 µl/well
  • Pre-blocked with SuperBlock™ buffer to reduce non specific binding