Pierce™ Protein A
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
29989 21181
PIER29989EA 196 EUR
PIER29989 PIER21181
Pierce™ Protein A
Eiwitzuivering Reagentia voor eiwitpurificatie
Protein A can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.
  • Contains four Fc-binding domains per protein
  • Efficient than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
  • Less efficient than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies