Anti-c-Myc magnetic beads, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88842 88843
PIER88842EA 477 EUR
PIER88842 PIER88843
Anti-c-Myc magnetic beads, Pierce™
Parels Magnetische parels
The Pierce™ Anti-c-Myc magnetic beads are affinity particles for immunoprecipitation of recombinant c-Myc-tagged proteins expressed in bacterial or mammalian cells or in vitro systems, using manual or robotic magnetic separators.
  • Highly specific anti-c-Myc monoclonal antibody enables high yield and high purity immunoprecipitation
  • Fast and convenient with low non-specific binding
  • Beads are compatible with manual and automated workflows
Stable, pre-blocked beads and specific antibody minimize off-target binding for c-Myc-tag IP or co-IP experiments