Pierce™ Microplates, Nickel Coated, 96 Well
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15142 15242 15342 15442
PIER15142EA 322 EUR
PIER15142 PIER15242 PIER15342 PIER15442
Pierce™ Microplates, Nickel Coated, 96 Well
Microplaten Immunologie-microplaten en -strips
These Ni(2+) chelate-coated plates are ideal for analysing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal. Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking. The clear, white or black plates can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively.
  • Ni2+ activated surface enables metal-chelate binding of His-tagged proteins
  • Detergents used to lyse cells don't inhibit binding to activated plates as they do with plain polystyrene
  • Better binding for sensitive assays compared to other commercially available nickel-activated plates
  • Detection limit: 1 ng of polyhistidine fusion protein
  • Binding capacity: approximately 9 pmol His-tagged protein (27 kDa) per well
  • Activation level: 200 µl