Microplates, copper coated, 96-well, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15143 15146 15147 15148
PIER15143EA 375 EUR
PIER15143 PIER15146 PIER15147 PIER15148
Microplates, copper coated, 96-well, Pierce™
Microplaten Immunologie-microplaten en -strips
Pierce Copper coated plates are ideal for analysing polyhistidine-tagged fusion proteins by ELISA-based methods. Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metals such as copper.
  • Quantitate previously undetectable histidine-tagged proteins
  • Wider dynamic range with four-fold greater capacity than regular nickel chelate-coated plate
  • Copper chelate provides greater binding capacity than nickel
  • Detection limit is 0,1 pg of polyhistidine tagged fusion protein per well
  • Pre-blocked for the ultimate convenience
These high binding capacity plates use an exclusive coating process to increase the amount of histidine-tagged protein that will bind to the plate surface.
Surface treatment: Divalent copper, metal chelate
Plate blocking: Blocker™ BSA buffer
Opgelet: Copper coated plates are for use with purified His-tagged proteins. Typical cell lysates contain non-his-tagged proteins that may bind to copper, which can reduce binding capacity and signal-to-noise ratios.