HyClone™ FetalClone™ II serum, U.S. Origin
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30066.03 SH30066.02 SH30066.02HI SH30066.03HI SH30066.03IR25
HYCLSH30066.03EA 352 EUR
HYCLSH30066.03 SH30066.02 HYCLSH30066.02HI HYCLSH30066.03HI SH30066.03IR25
HyClone™ FetalClone™ II serum, U.S. Origin
Serum
HyClone™ FetalClone® II is a foetal bovine serum alternative that contains additional growth factors and supplements in the formulation. It has been developed specifically for growth of CHO cells and derivatives.
  • IgG levels comparable with those found in FBS
  • Improved cell growth performance relative to FBS
  • High lot-to-lot consistency
  • Low endotoxin and hemoglobin levels
Sterile filtered using three sequential 100 nm (0,1 µm) pore size rated filters. Each lot undergoes extensive quality performance and analytical testing.