Affinity chromatography media, glutathione superflow agarose, pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25239 25238 25237 25236
PIER25239EA 9990 EUR
PIER25239 PIER25238 PIER25237 PIER25236
Affinity chromatography media, glutathione superflow agarose, pierce™
Eiwitzuivering Protein Purification Resins
Pierce™ Glutathione Superflow Agarose consists of glutathione immobilised to a crosslinked 6% beaded agarose for purification of GST-tagged proteins.
  • Binds 10 mg of pure GST per ml of resin at a linear flow rate of 150 cm/hour; can bind >20 mg/ml at 30 cm/hour flow rate
  • Provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
  • Highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200 cm/hour
  • Maintains function after exposure to a wide variety of chemicals and pH values
  • Resin is stable through multiple cycles of cleaning and reuse
Glutathione Superflow Agarose is a durable resin that does not compress at flow rates used for medium- or large-scale FPLC purifications.