Dialysis tubing, SnakeSkin™, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
68100 68035 68700 88242 88243 88244 88245
PIER68100EA 247 EUR
PIER68100 PIER68035 PIER68700 PIER88242 PIER88243 PIER88244 PIER88245
Dialysis tubing, SnakeSkin™, Pierce™
Slangen Dialyseslangen
SnakeSkin dialysis tubings with pleated format are made from high performance regenerated-cellulose membrane which has minimal nonspecific binding and a reliable molecular-weight cutoff (MWCO).
  • Ready-to-use, pre-opened and pre-wetted
  • Greater than 90% protein recovery
Regenerated cellulose is exceptionally stable when exposed to common laboratory solutions such as acidic and basic conditions, or hydrophobic solvents and alcohols. Ideal for dialysing protein samples.
Leveringsinformatie: Supplied in eight-inch (20 cm) cylinders containing 35 feet (10,7 m) of tubing.