Cell-free protein expression kits, 1-step human high yield mini IVT kit
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88890 88891
PIER88890EA 381 EUR
PIER88890 PIER88891
Cell-free protein expression kits, 1-step human high yield mini IVT kit
Eiwitexpressie Systemen voor eiwitexpressie
The Thermo Scientific 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per mL than other mammalian IVT systems.
  • High expression with up to 750 µg/ml of expressed protein
  • Low variability between experiments
  • Express high levels of protein with six hours to overnight incubation
  • Obtain functionally active proteins, including those containing disulfide bonds
  • Transcription and translation is performed in one reaction step
  • Amenable to incorporation of heavy or unnatural amino acids
The 1-Step Human High-Yield In Vitro Translation (IVT) Kits enable the expression of functional proteins with 10 to 100 times greater yield per ml than other mammalian IVT systems. The 1-Step High-Yield IVT System uses modified HeLa cell extracts to take advantage of the robust human translation machinery and generate functional full-length proteins. In this system, protein expression is performed in a proprietary dialysis device that allows a continuous supply of nucleotides, amino acids and energy generating substrates into the reaction while removing inhibitors of proteins synthesis. This continuous-exchange cell-free (CECF) system enables protein expression in an overnight incubation of up to 750 µg/ml. The complete mini-scale kit includes all the components required for transcription and translation of a recombinant gene, including an optimised expression vector.
Leveringsinformatie: Each 1- Step Human High-Yield Mini IVT Kit contains HeLa lysate (110 or 500 µl), accessory proteins (25 or 100 µl), reaction mix (40 or 200 µl), 5X dialysis buffer (600 µl or 3 ml), positive control DNA (pCFE-GFP - 10 µg), cloning vector (pT7CFE1-NHis-GST-CHA - 10 µg), microdialysis devices (2 or 10), and nuclease-free water (5 ml or 3×5 ml).
Opgelet: For research Use Only. Not for use in diagnostic procedures.