Spin purification kit for GST proteins, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16106 16107 16108
PIER16106EA 444 EUR
PIER16106 PIER16107 PIER16108
Spin purification kit for GST proteins, Pierce™
Eiwitzuivering Protein Purification Resins
Pierce™ GST Spin Purification Kit provides a fast, easy to use format for purifying GST-fusion proteins from cellular lysates.
  • Glutathione is immobilised through its central sulfhydryl onto 6% crosslinked agarose resin
  • High capacity binds at least 40 mg of purified recombinant GST protein per millilitre of resin
  • Consistently purifies at least 10 mg of GST-tagged protein per milliliter of resin with greater than 90% purity
  • Resin can be reused at least five times without reduction in performance
Leveringsinformatie: Kit includes Pierce™ Glutathione spin columns, Equilibration/Wash Buffer, 125 mM Tris, Glutathione, and column plugs. The 0,2 ml kit also includes collection tubes.