Pierce™ PEGylated SMCC Crosslinkers, Heterobifunctional
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22104 22102 22108 22112 22105 22107 22114 22113 22103
PIER22104EA 439 EUR
PIER22104 PIER22102 PIER22108 PIER22112 PIER22105 PIER22107 PIER22114 PIER22113 PIER22103
Pierce™ PEGylated SMCC Crosslinkers, Heterobifunctional
Koppelings- en labelingssystemen voor eiwitten
Cross linkers are used for the identification of near-neighbour protein relationships and ligand-receptor interactions. Homobifunctional, amine-reactive NHS esters or imidates, and heterobifunctional, amine-reactive, photoactivatable phenyl azides are the most commonly used crosslinkers for these applications.
Thermo Scientific Pierce SM(PEG)n is a series of amine-to-sulfhydryl crosslinkers that differ in length from 17,6 to 95,2 angstroms as a result of polyethylene glycol spacer arms containing n equals 2 to 24 ethylene glycol units.