HyClone AdvanceSTEM™ Stem Cell Media, Mesenchymal Stem Cell Expansion Kit, HyClone products (Cytiva)
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30878.01 SH30879.01 SH30878.02
HYCLSH30878.01EA 128 EUR
HYCLSH30878.01 HYCLSH30879.01 HYCLSH30878.02
HyClone AdvanceSTEM™ Stem Cell Media, Mesenchymal Stem Cell Expansion Kit, HyClone products (Cytiva)
Celcultuurmedia
AdvanceSTEM™ Mesenchymal Stem Cell Basal Medium and Mesenchymal Stem Cell Growth Supplement have been formulated to support expansion and maintenance of undifferentiated human mesenchymal stem cells (hMSCs).
  • Mycoplasma free and sterility tested
  • Stored at –10 °C or lower away from light
Supported cells include Human Wharton's Jelly Stem Cells (HWJSCs), Human Adipose-derived Mesenchymal Stem Cells (HAMSCs), Human Placental Mesenchymal Stem Cells (HPMSCs), Multipotent Cord Blood Unrestricted Somatic Stem Cells (MUCBUSSCs) and bone marrow-derived hMSCs.
It provides optimized conditions for embryonic stem cell expansion and differentiation.