Contrast clips
Leverancier: witeg Labortechnik
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3.691.001 3.691.002 3.691.003 3.691.004 3.691.005 3.691.006
WITG3.691.001EA 4 EUR
WITG3.691.001 WITG3.691.002 WITG3.691.003 WITG3.691.004 WITG3.691.005 WITG3.691.006
Contrast clips
Klemmen Klemmen voor algemeen gebruik
PP, white/black.
  • For attachment to all measuring flasks
  • With grip deepening
  • Unbreakable, resistant to acids and alkalis
  • For better recognition and adjustment of the liquid level