HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30118.03 SH30118.02 SH30118.04 SH30118.02HI SH30118.03HI SH30118.04HI SH30118.03IR25
HYCLSH30118.03EA 121 EUR
HYCLSH30118.03 HYCLSH30118.02 HYCLSH30118.04 HYCLSH30118.02HI HYCLSH30118.03HI SH30118.04HI SH30118.03IR25
HyClone™ Newborn Bovine Calf Serum, US Origin, Cytiva
Serum
Bovine calf serum products are excellent and cost-effective alternatives to fetal bovine serum (FBS). Bovine calf serum products contain exceptionally high levels of transferrin, which can provide three to four times as much available iron as FBS.
  • Complete traceability back to original source
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
HyClone™ Newborn Bovine Calf Serum, US Origin undergoes the same careful collection and processing procedures (including venipuncture) used for our bovine serum products. Newborn calf serum is sourced from animals that are typically less than 10 days old and filtered through three sequential 100 nm (0.1 μm) pore sized-rated filters.