Volumetric flasks for sugar analysis
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632431510030 632431510037
KAVA632431510030EA 66.1 EUR
KAVA632431510030 KAVA632431510037
Volumetric flasks for sugar analysis
Kolven Maatkolven
Borosilicate glass, clear.
  • With two graduation marks
  • White markings
  • Autoclavable at 121 °C