Kinase enrichment kits, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88310 88312
PIER88310EA 890 EUR
PIER88310 PIER88312
Kinase enrichment kits, Pierce™
Testen Enzyme-assays
The Pierce™ Kinase Enrichment Kits utilise an ActivX™ ATP or ADP probe to covalently label the active site of ATPases, including chaperones and metabolic enzymes, to enable their selective enrichment using a desthiobiotin tag.
  • Labels only the conserved active-site lysines of nucleotide-binding proteins
  • Flexible use for in vitro labeling of ATPase enzymes derived from cells or tissues
  • Compatible with use with Western blot or mass spectrometry (MS) workflows
ActivX™ probes feature an amine-reactive nucleotide analog and a desthiobiotin (biotin analogue) tag that facilitates selective labeling of lysines in the kinase active site and then subsequent enrichment and recovery of labelled protein.