Imject™ Bovine serum albumin (BSA), maleimide activated
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
77115 77116
PIER77115EA 554 EUR
PIER77115 PIER77116
Imject™ Bovine serum albumin (BSA), maleimide activated
Eiwitten en peptiden
Imject™ BSA maleimide-activated enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunisation and antibody production.