Hand cleansing cream, Solopol® classic
Leverancier: Deb-STOKO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SOL250ML SOL2LT SOL4LTR
115-2367EA 42.24 EUR
115-2367 115-2368 115-2370
Hand cleansing cream, Solopol® classic
Huidverzorging Zepen en huidreinigingsmiddelen
Traditional solvent-free hand cleanser paste with scrubbing agent for heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in industrial environments.
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • Contains Eucornol® skin protectant to minimise the risk of skin irritation
  • Solvent-free, colourant-free, perfumed
Bestelinformatie: Soft bottle may only be used with Stoko® Vario® dispensers.