SFM4 insect media, HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30913.02 SH30912.01 SH30913.03
SH30913.02EA 80.9 EUR
SH30913.02 SH30912.01 SH30913.03
SFM4 insect media, HyClone™
Celcultuurmedia
HyClone™ SFM4 insect is a versatile cell culture medium developed through Metabolic Pathway Design to support the growth of multiple insect cell lines and production of a variety of recombinant proteins without compromising performance. HyClone SFM4 insect provides superior growth of many key insect cell lines, including Sf9, Sf21, High Five (T.ni), and D.mel.
  • Supports multiple insect cell lines
  • Free of animal derived components
  • Protein-free
  • High performance growth in multiple culture systems, including high density bioreactors
  • Full component traceability
  • Manufactured in cGMP and ISO compliant facility
Certificaten: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).