Dialysis flasks, Slide-A-Lyzer™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
87761 87762 87760 87763
PIER87761EA 412 EUR
PIER87761 PIER87762 PIER87760 PIER87763
Dialysis flasks, Slide-A-Lyzer™
Slangen Dialyseslangen
These dialysis flasks facilitate simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules in sample volumes up to 250 ml.
  • Easy to use
  • Color-coded for easy identification
  • No leakage, sample remains intact with no contamination
  • High surface area to volume ratio maximizes diffusion rate
Devices are constructed from two sheets of low protein-binding, regenerated cellulose membranes to ensure maximum sample recovery and purity and contain up to 85% less plastic per volume compared to other devices. Slide-A-More Product Information