Insect cell culture media, serum-free, CCM3, HyClone™
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30065.02 SH30061.03
HYCLSH30065.02EA 77.7 EUR
HYCLSH30065.02 HYCLSH30061.03
Insect cell culture media, serum-free, CCM3, HyClone™
Celcultuurmedia
HyClone™ CCM3 media is designed to increase cell yields with multiple lines in a variety of culture environments. It is tailored for Sf9 cells.
The cell culture media is developed through metabolic pathway design to support superior performance and increased process yields of multiple cell culture platforms. The serum free media have been designed to provide a consistent nutrient supply without the need for serum prescreening.