HyClone™ ADCF-mAb media
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30349.02 SH30349.LS SH30635.01
HYCLSH30349.02EA 75.4 EUR
HYCLSH30349.02 HYCLSH30349.LS HYCLSH30635.01
HyClone™ ADCF-mAb media
Celcultuurmedia
Protein-free and animal-derived component-free medium
  • Animal-derived component-free (ADCF) formulation
  • Designed for high cell yield and monoclonal antibody (MAb) production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
ADCF-mAb liquid medium contains poloxamer 188 and L-glutamine, and does not contain phenol red. Liquid medium is also available without L-glutamine to support the glutamine synthetase (GS) gene expression system. ADCF‑mAb powder medium does not contain poloxamer 188, L-glutamine, phenol red, or sodium bicarbonate. ADCF-mAb is available in liquid and powder formats in user-friendly packaging.
HyClone ADCF-mAb cell culture medium is protein-free and contains no animal-derived components. This regulatory-friendly medium is developed to increase the process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.