Western blot enhancer, SuperSignal™, Pierce
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
46640 46641
PIER46640EA 348 EUR
PIER46640 PIER46641
Western blot enhancer, SuperSignal™, Pierce
Reagentia voor elektroforese Western Blotting Reagents
SuperSignal™ Western Blot Enhancer is a membrane treatment and a primary antibody diluent that has been shown to increase intensity of a target protein band on a Western blot and to enable detection of a target analyte at a level at which it could not be detected without enhancement.
  • Increase in signal intensity and sensitivity
  • Improves signal-to-noise ratio for poor quality and low affinity antibodies
  • Provides effective signal enhancement with PVDF and nitrocellulose membranes
  • Ready-to-use and can be stored at room temperature