HyClone™ SFM4MAb media
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30513.02 SH30535.02
HYCLSH30513.02EA 76.1 EUR
HYCLSH30513.02 HYCLSH30535.02
HyClone™ SFM4MAb media
Celcultuurmedia
HyClone™ SFM4MAb media was developed through the Metabolic Pathway Design approach to increase process yields for manufacturing of human and humanised recombinant antibodies for therapeutic use.
Order Now
Specificaties
Environmentally Preferable