Pierce™ Immunoassay Microplates, Protein A, G, A/G Coated Plates
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15131 15132 15130 15133 15138 15154 15155 15156 15157
PIER15131EA 314 EUR
PIER15131 PIER15132 PIER15130 PIER15133 PIER15138 PIER15154 PIER15155 PIER15156 PIER15157
Pierce™ Immunoassay Microplates, Protein A, G, A/G Coated Plates
Microplaten Immunologie-microplaten en -strips
Pierce™ Protein A, G and A/G coated plates provide an alternative to direct, passive adsorption methods for immobilising antibodies for ELISA and other plate-based assay techniques. Available in clear for colorimetric assays, white for chemiluminescent assays, and black for fluorescent assays.
  • Retain antibody activity, which can be lost when antibodies are immobilised by passive adsorption
  • Orient antibodies for maximum antigen-binding capacity
  • Immobilise antibodies for plate assays without prior purification
  • Ensure minimal variation (<5% well-to-well)