HyClone™ SFM4CHO media
Leverancier: HyClone products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30549.02 SH30548.02 SH30548.LS SH30518.01 SH30549.LS
HYCLSH30549.02EA 75.9 EUR
HYCLSH30549.02 HYCLSH30548.02 HYCLSH30548.LS HYCLSH30518.01 HYCLSH30549.LS
HyClone™ SFM4CHO media
Celcultuurmedia
HyClone™ SFM4CHO media is a serum-free and protein-free formulation that contains no components of bovine origin and has been designed for high performance in a variety of culture vessels, including bioreactors.
  • Metabolically designed for high cell yield and recombinant protein production
  • Supports multiple CHO clones
  • Allows for direct or sequential adaptation
  • Supports growth in multiple cell culture systems
  • Component traceability
  • Manufactured under cGMP
HyClone™ SFM4CHO media has been formulated using a proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines. It has been designed to support the DHFR selection/amplification system and is also available without L-glutamine to support the GS gene expression system.