Immobilised NeutrAvidin™, UltraLink™, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
53151 53150
PIER53151EA 558 EUR
PIER53151 PIER53150
Immobilised NeutrAvidin™, UltraLink™, Pierce™
Eiwitzuivering Protein Purification Resins
NeutrAvidin™ UltraLink™ resin is a beaded polyacrylamide resin of immobilised NeutrAvidin™ Protein, a modified form of avidin with exceptional biotin-binding characteristics for affinity purification methods.
  • UltraLink beads are hydrophilic, charge-free, highly crosslinked, rigid and durable
  • Acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium-pressure applications
NeutrAvidin™ UltraLink™ resin consists of a specially purified and deglycosylated form of avidin that has been covalently immobilised onto a beaded acrylamide support. Compared to native avidin, NeutrAvidin™ Protein is more neutral and exhibits much less non specific binding properties in biotin-binding applications. UltraLink beads are hydrophilic, charge-free, highly crosslinked, rigid and durable. These properties make the affinity resin suitable for a variety of batch- or column-type affinity procedures that require medium-pressure chromatography.
NeutrAvidin™ UltraLink™ resin binding capacity: 12 - 20 µg biotin/ml of settled resin
NeutrAvidin™ Plus UltraLink™ resin binding capacity: >30 µg biotin/ml of settled resin