Immunoassay microplates, maleimide activated, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
15150 15152 15153
PIER15150EA 225 EUR
PIER15150 PIER15152 PIER15153
Immunoassay microplates, maleimide activated, Pierce™
Microplaten Immunologie-microplaten en -strips
Pierce™ Maleimide activated plates are ideal for binding sulfhydryl-containing molecules that are difficult to coat onto polystyrene plates, such as peptides that contain a terminal cysteine. Available in clear for colorimetric assays, white for chemiluminescent assays and black for fluorescent assays.
  • Pre-blocked bovine serum albumin (BSA) to reduce non specific binding
  • Binding capacity is 100 to 150 pmol of a sulfhydryl-containing peptide (307 kDa) per well
  • Activation level: 100 µl
Pierce™ Maleimide activated plates provide covalent attachment (More Product Information