Peroxidase conjugate stabiliser/diluent, Guardian™
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
37548
37552
PIER37548EA
168
EUR
PIER37548
PIER37552
Peroxidase conjugate stabiliser/diluent, Guardian™
Reagentia voor elektroforese
Western Blotting Reagents
Guardian™ Peroxidase conjugate stabiliser/diluent preserves the functional integrity and activity of horseradish peroxidase conjugated antibodies and other proteins at very dilute concentrations for long term storage.
- Preserves HRP activity – no significant loss of HRP activity over a six-month period at room temperature (1:1000 dilution) or 12 months at 4 °C
- Store ready-to-use dilutions (1:1000 to 1:100,000) that maintain enzyme activity in the refrigerator – no aliquoting or freezing
- Assay compatible – simply add blocking buffer to create the ideal diluent for HRP-based ELISA systems or store the HRP conjugate as a 1:1000 stock solution for Western blots and dilute in the final assay buffer