HyClone™ SFM4HEK293, HEK 293 Media
Leverancier: HyClone products (Cytiva)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SH30521.02
SH30522.01
HYCLSH30521.02EA
86.3
EUR
HYCLSH30521.02
HYCLSH30522.01
HyClone™ SFM4HEK293, HEK 293 Media
Celcultuurmedia
HyClone™ SFM4HEK293 is a protein-free cell culture media without animal-derived components (ADCF) used to support production of adenoviral vectors and recombinant proteins in HEK 293 cultures.
- Developed through metabolic pathway design process for high adenoviral vector and protein production
- Ready-to-use liquid formulation requires no supplementation
- Allows for direct or sequential adaptation
- For large-scale culture applications, including perfusion and fed-batch strategies
- Manufactured according to cGMP guidelines