Protein A and Protein G spin plates for IgG screening, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45202 45204
PIER45202EA 366 EUR
PIER45202 PIER45204
Protein A and Protein G spin plates for IgG screening, Pierce™
Microplaten Immunologie-microplaten en -strips
Pierce™ Protein A and Protein G Spin Plates for IgG Screening offers a high throughput format for quick purification and screening of antibodies and for immunoprecipitation (IP). Each well of the 96-well spin plate can process 10 to 100 µl of serum, cell culture supernatant or ascites fluid samples.
The Protein A and Protein G resin purifies by binding IgGs, primarily through their Fc region, in a buffer that facilitates the interaction. Non-relevant proteins, such as albumin and transferrin, are removed by washing and the purified antibody is then recovered using low-pH elution conditions.
Leveringsinformatie: Each pack contains 1 plate (each well contains 50 µl of resin for processing 10 to 100 µl of sample) and 3 wash/collection plates.