Anti-c-Myc agarose, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20168 20169
PIER20168EA 698 EUR
PIER20168 PIER20169
Anti-c-Myc agarose, Pierce™
Eiwitzuivering Protein Purification Resins
Pierce™ Anti-c-Myc Agarose is a high-affinity immunoprecipitation resin ideal for purification of recombinant c-Myc-tagged proteins expressed in human in vitro expression systems and bacterial and mammalian cell lysates.
  • Highly specific anti-c-Myc monoclonal antibody (clone 9E10) enables high yield and high purity for immunoprecipitation
  • Available in 2 and 10 mL resin package sizes to allow for larger scale purifications or immunoprecipitations
  • Can be used in spin or gravity columns as well as in FPLC cartridges
Order Now
Specificaties
Environmentally Preferable