Desalting columns, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
43230 43233
PIER43230EA 160 EUR
PIER43230 PIER43233
Desalting columns, Pierce™
Kolommen voor chromatografie
These desalting columns are ready-to-use, disposable, gel filtration columns for separating proteins and other macromolecules from low molecular weight buffer salts and reagents.
  • Stable in water, salt solutions, organic solvents and alkaline or acidic conditions
  • Excellent flow properties
  • Heat stable
  • Gravity flow column format
  • Purification targeting buffer exchange, protein
  • Disposable
They have wide range of applications that includes removing salts from protein solutions, removing phenol from nucleic acid preparations, separating excess crosslinker from conjugate preparations, removing excess derivatising agents from modified proteins, removing unreacted dye from fluorescent antibodies, removing free radiolabel from labeled proteins and exchanging one buffer for another.
Gravity-flow gel filtration involves the chromatographic separation of molecules of different dimensions based on their relative abilities to penetrate a suitable stationary phase. A chromatographic matrix, usually consisting of very small, uncharged porous particles in an aqueous solution, is packed into a column and then used for separation via size exclusion.
Different levels of separation can be achieved based on the pore size of the medium packed into the desalting column-in our case, dextran. With size exclusion filtration, large molecules are excluded from the internal pores of the gel and emerge from the column first. Smaller molecules can penetrate the pores, then progress through the column at a slower rate. These smaller molecules are subsequently flushed through the column with additional buffer volume.
Dextran is stable in water, salt solutions, organic solvents and alkaline or weakly acidic solutions. Dextran is also heat-stable and can be autoclaved dry or in solution at a neutral pH for 30 minutes at 120 °C without affecting its chromatographic properties.