Ladies fleece jackets, Acode® BaseCamp 1498
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
119011940XL 119011331S 119011331M 119011331L 119011331XL 1190113312XL 119011540S 119011540M 119011540L 119011540XL 1190115402XL 119011930S 119011930M 119011930L 119011930XL 1190119302XL 119011940S 119011940M 119011940L 1190119402XL
KWIN119011940XLEA 43.9 EUR
KWIN119011940XL KWIN119011331S KWIN119011331M KWIN119011331L KWIN119011331XL KWIN1190113312XL KWIN119011540S KWIN119011540M KWIN119011540L KWIN119011540XL KWIN1190115402XL KWIN119011930S KWIN119011930M KWIN119011930L KWIN119011930XL KWIN1190119302XL KWIN119011940S KWIN119011940M KWIN119011940L KWIN1190119402XL
Ladies fleece jackets, Acode® BaseCamp 1498
Werkkleding
Comfortable fleece jacket made of 100% polyester fleece.
  • Flat decorative seams
  • Side pockets with zip
  • Slightly tailored, extended back
  • Black elastic edging at neck and cuffs
  • Elbow patches in tone-in-tone fleece
  • Drawstring hem
Weight: 280 g/m²