Visuele kleurenkits, chromaat, MColortest™, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1.14402.0001 1.14441.0001 1.14756.0001
1.14402.0001EA 173 EUR
1.14402.0001 1.14441.0001 1.14756.0001
Visuele kleurenkits, chromaat, MColortest™, Supelco®
Milieutestkits
Snelle tests, visuele kleurvergelijking.
  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable
  • A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
MColortest™ met kleurschijfcomparator (voorheen Microquant®)
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
Leveringsinformatie: Elke testkit bevat alle benodigde reagentia, reactiecuvetten, een kleurvergelijkingskaart of kleurschijf en uitgebreide gebruiksinstructies.