Microdialysis plates, 96-well, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
88260 88262
PIER88260EA 610 EUR
PIER88260 PIER88262
Microdialysis plates, 96-well, Pierce™
Microplaten Deepwell-microplaten
Pierce™ 96-well microdialysis plates are automation-compatible microplate dialysis devices suitable for rapidly processing 1 to 96 samples with volumes 10 to 100 µl.
  • High yield, > 90% protein recovery after dialysis
  • Dialysis completed in 2 to 4 hours
  • Sample loading and retrieval can be performed using a standard pipette
  • Detachable 8-device strips, scalable from 1 to 96 samples
  • Plate format conforms to the SBS microplate standard
Pierce™ 96-well microdialysis plates are suitable for applications such as protein and DNA desalting/buffer exchange, removal of dyes after protein labeling, enzyme activity/binding assays, and drug binding studies.
Each sample processed with the 96-well microdialysis dialysis plate is dialysed inside a single, removable dialysis device created by two regenerated cellulose membranes separated by ~2 mm. The short diffusion distance and large surface area facilitates rapid dialysis with the removal of salts and small molecule. Sample recovery is performed using standard laboratory micropipettes. Individual microdialysis devices can also be used with 2 ml microcentrifuge tubes for processing single samples. The available plate seals are made of high-quality polypropylene and help protect samples from contamination and evaporation, especially during long-term storage.
Leveringsinformatie: Kit includes 12 cartridges of eight microdialysis devices in a 96 deep well plate.