Pegylation reagents, Methyl-PEG-NHS-Maleimide, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22711 22713
PIER22711EA 476 EUR
PIER22711 PIER22713
Pegylation reagents, Methyl-PEG-NHS-Maleimide, Pierce™
Eiwitzuivering Reagentia voor eiwitpurificatie
Pierce™ MM(PEG)n reagents are methyl-terminated PEG compounds activated with a maleimide group for covalent pegylation of sulfhydryls on proteins or assay surfaces.
  • Choice of 12 and 24 ethylene glycol units, enabling modification procedures to be optimised for a specific application
  • PEG spacer provides advantages that include increased stability, reduced tendency toward aggregation and reduced immunogenicity
  • Maleimide activated for efficient PEGylation of primary amines at pH 6,5 to 7,5; reaction of maleimide group results in formation of stable, irreversible amide bonds
  • Easy to follow instructions increase the likelihood of a successful outcome