Recombinant protein A agarose, Pierce™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
20365 20366
PIER20365EA 466 EUR
PIER20365 PIER20366
Recombinant protein A agarose, Pierce™
Eiwitzuivering Protein Purification Resins
Pierce™ Recombinant Protein A agarose is a beaded agarose resin to which a genetically engineered form of Protein A has been immobilised for use in IgG purification applications where the native ligand has undesirable effects.
  • Immobilised Protein A is ideal for polyclonal IgG purification from human and rabbit
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Standard capacity beads with a normal load of immobilized Protein A, providing a binding capacity of 15 to 17 mg human IgG/ml resin
Although some non essential regions have been removed, recombinant is similar to native Protein A in overall size and IgG-binding domains. The protein has four high affinity binding sites capable of specifically binding to the Fc region of immunoglobulin molecules from several species.