Pierce™ Vacuum Hydrolysis Tubes
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
29570 29571 29572
PIER29570EA 252 EUR
PIER29570 PIER29571 PIER29572
Pierce™ Vacuum Hydrolysis Tubes
Buisjes Reageerbuisjes
These are high quality glass tubes designed for heated, acid-based, vacuum hydrolysis of proteins and peptides for amino acid analysis. It fits conveniently into reacti-block aluminium heating blocks.
  • Fits into Reacti-Block aluminium heating blocks
  • Sufficient for samples up to 0,35 ml
  • Optimal hydrolysis control
The upper temperature limit of the vacuum hydrolysis tubes is 260 °C, however, do not heat the tubes greater than 100 °C in an oven.